Assay Details
| Gene Symbol: CHR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335605.1 | NP_001318246.1 | 4 - 5 | 1164 | 73 | |
| NM_001335606.1 | NP_001324143.1 | 4 - 5 | 1215 | 73 | ||
| NM_001335608.1 | NP_001324146.1 | 4 - 5 | 1215 | 73 | ||
| NM_001335609.1 | NP_001324145.1 | 4 - 5 | 1237 | 73 | ||
| NM_001335610.1 | NP_001324144.1 | 3 - 4 | 1392 | 73 | ||
| NM_127432.2 | NP_179466.1 | 3 - 4 | 1392 | 73 | ||
More Information
Set Membership: |
|

CHR8
Genomic Map