Assay Details
Gene Symbol: SLP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335625.1 | NP_001324171.1 | 9 - 10 | 1943 | 75 | |
NM_127474.4 | NP_565447.1 | 9 - 10 | 2065 | 75 | ||
GenBank mRNA | AY051009.1 | - | 9 - 10 | 2091 | 75 | |
AY133826.1 | - | 8 - 9 | 1756 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
