Assay Details
| Gene Symbol: FRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127476.2 | NP_179509.1 | 2 - 3 | 681 | 102 | |
| GenBank mRNA | AF486619.1 | - | 2 - 3 | 681 | 102 | |
| FJ708695.1 | - | 2 - 3 | 630 | 102 | ||
More Information
Set Membership: |
|

FRK1
Genomic Map