Assay Details
| Gene Symbol: TAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127503.3 | NP_179535.1 | 12 - 13 | 1617 | 101 | |
| GenBank mRNA | AJ131831.1 | - | 12 - 13 | 1528 | 101 | |
| AJ238008.1 | - | 12 - 13 | 1436 | 101 | ||
| AY054480.1 | - | 12 - 13 | 1459 | 101 | ||
| BT008883.1 | - | 12 - 13 | 1298 | 101 | ||
More Information
Set Membership: |
|

TAG1
Genomic Map