Assay Details
| Gene Symbol: TAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127503.3 | NP_179535.1 | 4 - 5 | 944 | 101 | |
| GenBank mRNA | AF051849.1 | - | 4 - 5 | 712 | 101 | |
| AJ131831.1 | - | 4 - 5 | 855 | 101 | ||
| AJ238008.1 | - | 4 - 5 | 763 | 101 | ||
| AY054480.1 | - | 4 - 5 | 786 | 101 | ||
| BT008883.1 | - | 4 - 5 | 625 | 101 | ||
More Information
Set Membership: |
|

TAG1
Genomic Map