Assay Details
| Gene Symbol: TAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127503.3 | NP_179535.1 | 9 - 10 | 1451 | 68 | |
| GenBank mRNA | AF051849.1 | - | 9 - 10 | 1219 | 68 | |
| AJ131831.1 | - | 9 - 10 | 1362 | 68 | ||
| AJ238008.1 | - | 9 - 10 | 1270 | 68 | ||
| AY054480.1 | - | 9 - 10 | 1293 | 68 | ||
| BT008883.1 | - | 9 - 10 | 1132 | 68 | ||
More Information
Set Membership: |
|

TAG1
Genomic Map