Assay Details
| Gene Symbol: AT2G19790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127537.4 | NP_179569.1 | 2 - 3 | 355 | 94 | |
| GenBank mRNA | AF370185.1 | - | 2 - 3 | 297 | 94 | |
| AY059143.1 | - | 2 - 3 | 228 | 94 | ||
| AY088273.1 | - | 2 - 3 | 299 | 94 | ||
More Information
Set Membership: |
|

AT2G19790
Genomic Map