Assay Details
| Gene Symbol: NADP-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335646.1 | NP_001324890.1 | 1 - 2 | 102 | 133 | |
| NM_127548.4 | NP_179580.1 | 3 - 4 | 475 | 133 | ||
| GenBank mRNA | AY062734.1 | - | 3 - 4 | 369 | 133 | |
| BT003371.1 | - | 3 - 4 | 319 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001335646.1 transcripts.
Set Membership: |
|

NADP-ME1
Genomic Map