Assay Details
| Gene Symbol: NADP-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335646.1 | NP_001324890.1 | 4 - 5 | 423 | 114 | |
| NM_127548.4 | NP_179580.1 | 6 - 7 | 796 | 114 | ||
| GenBank mRNA | AY062734.1 | - | 6 - 7 | 690 | 114 | |
| BT003371.1 | - | 6 - 7 | 640 | 114 | ||
More Information
Set Membership: |
|

NADP-ME1
Genomic Map