Assay Details
| Gene Symbol: NADP-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335646.1 | NP_001324890.1 | 5 - 6 | 522 | 151 | |
| NM_127548.4 | NP_179580.1 | 7 - 8 | 895 | 151 | ||
| GenBank mRNA | AY062734.1 | - | 7 - 8 | 789 | 151 | |
| BT003371.1 | - | 7 - 8 | 739 | 151 | ||
More Information
Set Membership: |
|

NADP-ME1
Genomic Map