Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202627.2 | NP_001189556.1 | 12 - 13 | 2071 | 59 | |
| NM_127553.3 | NP_179585.3 | 12 - 13 | 1693 | 59 | ||
| GenBank mRNA | AK229460.1 | - | 12 - 13 | 1693 | 59 | |
| BT011678.1 | - | 12 - 13 | 1507 | 59 | ||
More Information
Set Membership: |
|

GC1
Genomic Map