Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202627.2 | NP_001189556.1 | 19 - 20 | 2588 | 82 | |
| NM_127553.3 | NP_179585.3 | 19 - 20 | 2210 | 82 | ||
| GenBank mRNA | AK229460.1 | - | 19 - 20 | 2210 | 82 | |
| BT011678.1 | - | 19 - 20 | 2024 | 82 | ||
More Information
Set Membership: |
|

GC1
Genomic Map