Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127553.3 | NP_179585.3 | 3 - 4 | 335 | 99 | |
| GenBank mRNA | AK229460.1 | - | 3 - 4 | 335 | 99 | |
| BT011678.1 | - | 3 - 4 | 149 | 99 | ||
More Information
Set Membership: |
|

GC1
Genomic Map