Assay Details
| Gene Symbol: CAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127560.4 | NP_565462.1 | 3 - 4 | 1244 | 102 | |
| GenBank mRNA | AY045882.1 | - | 3 - 4 | 1210 | 102 | |
| BT004337.1 | - | 3 - 4 | 1149 | 102 | ||
More Information
Set Membership: |
|

CAF1
Genomic Map