Assay Details
| Gene Symbol: CAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127560.4 | NP_565462.1 | 4 - 5 | 1371 | 114 | |
| GenBank mRNA | AY045882.1 | - | 4 - 5 | 1337 | 114 | |
| BT004337.1 | - | 4 - 5 | 1276 | 114 | ||
More Information
Set Membership: |
|

CAF1
Genomic Map