Assay Details
| Gene Symbol: PIL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202630.2 | NP_001189559.1 | 2 - 3 | 334 | 124 | |
| NM_001335663.1 | NP_001318252.1 | 2 - 3 | 329 | 124 | ||
| NM_001335664.1 | NP_001323902.1 | 1 - 2 | 461 | 124 | ||
| NM_001335666.1 | NP_001323904.1 | 1 - 2 | 565 | 124 | ||
| NM_001335668.1 | NP_001323901.1 | 1 - 2 | 461 | 124 | ||
| GenBank mRNA | AF488560.2 | - | 1 - 2 | 99 | 124 | |
More Information
Set Membership: |
|

PIL5
Genomic Map