Assay Details
| Gene Symbol: PIL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202630.2 | NP_001189559.1 | 5 - 6 | 1180 | 135 | |
| NM_001335663.1 | NP_001318252.1 | 5 - 6 | 1175 | 135 | ||
| NM_001335664.1 | NP_001323902.1 | 4 - 5 | 1307 | 135 | ||
| NM_001335665.1 | NP_001323903.1 | 4 - 5 | 1270 | 135 | ||
| NM_001335666.1 | NP_001323904.1 | 4 - 5 | 1411 | 135 | ||
| NM_001335667.1 | NP_001323905.1 | 4 - 5 | 1374 | 135 | ||
| NM_179665.3 | NP_849996.1 | 5 - 6 | 1138 | 135 | ||
| GenBank mRNA | AF488560.2 | - | 4 - 5 | 945 | 135 | |
| BT029775.1 | - | 3 - 4 | 732 | 135 | ||
More Information
Set Membership: |
|

PIL5
Genomic Map