Assay Details
| Gene Symbol: PIL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335663.1 | NP_001318252.1 | 7 - 8 | 1577 | 88 | |
| NM_001335666.1 | NP_001323904.1 | 6 - 7 | 1813 | 88 | ||
| NM_001335667.1 | NP_001323905.1 | 6 - 7 | 1776 | 88 | ||
| NM_179665.3 | NP_849996.1 | 7 - 8 | 1540 | 88 | ||
| GenBank mRNA | AF488560.2 | - | 6 - 7 | 1347 | 88 | |
| BT029775.1 | - | 5 - 6 | 1134 | 88 | ||
More Information
Set Membership: |
|

PIL5
Genomic Map