Assay Details
| Gene Symbol: PIL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335663.1 | NP_001318252.1 | 8 - 9 | 1746 | 109 | |
| NM_179665.3 | NP_849996.1 | 8 - 9 | 1709 | 109 | ||
| GenBank mRNA | AF488560.2 | - | 7 - 8 | 1516 | 109 | |
More Information
Set Membership: |
|

PIL5
Genomic Map