Assay Details
| Gene Symbol: RIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127602.4 | NP_179633.2 | 2 - 3 | 596 | 72 | |
| GenBank mRNA | DQ446531.1 | - | 2 - 3 | 177 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIC6| Gene Symbol: RIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127602.4 | NP_179633.2 | 2 - 3 | 596 | 72 | |
| GenBank mRNA | DQ446531.1 | - | 2 - 3 | 177 | 72 | |
Set Membership: |
|