Assay Details
| Gene Symbol: TIM44-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335690.1 | NP_001325326.1 | 10 - 11 | 1275 | 85 | |
| NM_001335691.1 | NP_001325325.1 | 10 - 11 | 1266 | 85 | ||
| NM_001335692.1 | NP_001325324.1 | 10 - 11 | 1157 | 85 | ||
| NM_127610.1 | NP_565473.1 | 10 - 11 | 1157 | 85 | ||
| GenBank mRNA | DQ446533.1 | - | 10 - 11 | 1163 | 85 | |
More Information
Set Membership: |
|

TIM44-1
Genomic Map