Assay Details
| Gene Symbol: RPN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127618.4 | NP_565477.1 | 11 - 12 | 1403 | 75 | |
| GenBank mRNA | AF360216.1 | - | 11 - 12 | 1355 | 75 | |
| AY040061.1 | - | 11 - 12 | 1154 | 75 | ||
| AY230828.1 | - | 11 - 12 | 1238 | 75 | ||
More Information
Set Membership: |
|

RPN1A
Genomic Map