Assay Details
| Gene Symbol: RPN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127618.4 | NP_565477.1 | 14 - 15 | 1751 | 76 | |
| GenBank mRNA | AF360216.1 | - | 14 - 15 | 1703 | 76 | |
| AY040061.1 | - | 14 - 15 | 1502 | 76 | ||
| AY230828.1 | - | 14 - 15 | 1586 | 76 | ||
More Information
Set Membership: |
|

RPN1A
Genomic Map