Assay Details
| Gene Symbol: RPN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127618.4 | NP_565477.1 | 18 - 19 | 2369 | 60 | |
| GenBank mRNA | AF360216.1 | - | 18 - 19 | 2321 | 60 | |
| AY040061.1 | - | 18 - 19 | 2120 | 60 | ||
| AY230828.1 | - | 18 - 19 | 2204 | 60 | ||
More Information
Set Membership: |
|

RPN1A
Genomic Map