Assay Details
| Gene Symbol: RPN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127618.4 | NP_565477.1 | 4 - 5 | 644 | 74 | |
| GenBank mRNA | AF360216.1 | - | 4 - 5 | 596 | 74 | |
| AY040061.1 | - | 4 - 5 | 395 | 74 | ||
| AY230828.1 | - | 4 - 5 | 479 | 74 | ||
More Information
Set Membership: |
|

RPN1A
Genomic Map