Assay Details
| Gene Symbol: SRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335713.1 | NP_001324598.1 | 8 - 9 | 920 | 106 | |
| NM_127654.4 | NP_565489.2 | 8 - 9 | 920 | 106 | ||
More Information
Set Membership: |
|

SRF1
Genomic Map