Assay Details
| Gene Symbol: LIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335714.1 | NP_001318257.1 | 3 - 4 | 1075 | 69 | |
| NM_001335715.1 | NP_001323612.1 | 3 - 4 | 1066 | 69 | ||
| NM_127655.4 | NP_179682.1 | 3 - 4 | 1091 | 69 | ||
| GenBank mRNA | AB007987.1 | - | 3 - 4 | 1066 | 69 | |
| AF424583.1 | - | 3 - 4 | 1034 | 69 | ||
| AY035143.1 | - | 3 - 4 | 1040 | 69 | ||
| AY059077.1 | - | 3 - 4 | 1009 | 69 | ||
More Information
Set Membership: |
|

LIP1
Genomic Map