Assay Details
| Gene Symbol: AT2G20930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127662.4 | NP_565493.1 | 2 - 3 | 311 | 72 | |
| GenBank mRNA | AK317476.1 | - | 2 - 3 | 176 | 72 | |
| BT025606.1 | - | 2 - 3 | 130 | 72 | ||
More Information
Set Membership: |
|

AT2G20930
Genomic Map