Assay Details
| Gene Symbol: SYTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202636.1 | NP_001189565.1 | 2 - 3 | 483 | 89 | |
| NM_001202637.1 | NP_001189566.1 | 2 - 3 | 483 | 89 | ||
| NM_127668.4 | NP_565495.1 | 2 - 3 | 565 | 89 | ||
| GenBank mRNA | AB102951.1 | - | 2 - 3 | 208 | 89 | |
| AJ617630.1 | - | 2 - 3 | 461 | 89 | ||
| AY045836.1 | - | 2 - 3 | 468 | 89 | ||
| AY087925.1 | - | 2 - 3 | 463 | 89 | ||
| BT004371.1 | - | 2 - 3 | 208 | 89 | ||
More Information
Set Membership: |
|

SYTA
Genomic Map