Assay Details
| Gene Symbol: SYTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202636.1 | NP_001189565.1 | 3 - 4 | 536 | 84 | |
| NM_001202637.1 | NP_001189566.1 | 3 - 4 | 536 | 84 | ||
| NM_127668.4 | NP_565495.1 | 3 - 4 | 618 | 84 | ||
| GenBank mRNA | AB102951.1 | - | 3 - 4 | 261 | 84 | |
| AJ617630.1 | - | 3 - 4 | 514 | 84 | ||
| AY045836.1 | - | 3 - 4 | 521 | 84 | ||
| AY087925.1 | - | 3 - 4 | 516 | 84 | ||
| BT004371.1 | - | 3 - 4 | 261 | 84 | ||
More Information
Set Membership: |
|

SYTA
Genomic Map