Assay Details
| Gene Symbol: SYTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202636.1 | NP_001189565.1 | 4 - 5 | 667 | 154 | |
| NM_001202637.1 | NP_001189566.1 | 4 - 5 | 667 | 154 | ||
| NM_127668.4 | NP_565495.1 | 4 - 5 | 749 | 154 | ||
| GenBank mRNA | AB102951.1 | - | 4 - 5 | 392 | 154 | |
| AJ617630.1 | - | 4 - 5 | 645 | 154 | ||
| AY045836.1 | - | 4 - 5 | 652 | 154 | ||
| AY087925.1 | - | 4 - 5 | 647 | 154 | ||
| BT004371.1 | - | 4 - 5 | 392 | 154 | ||
More Information
Set Membership: |
|

SYTA
Genomic Map