Assay Details
| Gene Symbol: SYTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202636.1 | NP_001189565.1 | 7 - 8 | 951 | 62 | |
| NM_127668.4 | NP_565495.1 | 7 - 8 | 1033 | 62 | ||
| GenBank mRNA | AB102951.1 | - | 7 - 8 | 676 | 62 | |
| AJ617630.1 | - | 7 - 8 | 929 | 62 | ||
| AY045836.1 | - | 7 - 8 | 936 | 62 | ||
| BT004371.1 | - | 7 - 8 | 676 | 62 | ||
More Information
Set Membership: |
|

SYTA
Genomic Map