Assay Details
| Gene Symbol: SYTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202637.1 | NP_001189566.1 | 10 - 11 | 1392 | 104 | |
| NM_127668.4 | NP_565495.1 | 9 - 10 | 1360 | 104 | ||
| GenBank mRNA | AB102951.1 | - | 9 - 10 | 1003 | 104 | |
| AJ617630.1 | - | 9 - 10 | 1256 | 104 | ||
| AY045836.1 | - | 9 - 10 | 1263 | 104 | ||
| AY087925.1 | - | 9 - 10 | 1258 | 104 | ||
| BT004371.1 | - | 9 - 10 | 1003 | 104 | ||
More Information
Set Membership: |
|

SYTA
Genomic Map