Assay Details
| Gene Symbol: PRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127684.4 | NP_179710.1 | 1 - 2 | 224 | 81 | |
| GenBank mRNA | AK117333.1 | - | 1 - 2 | 216 | 81 | |
| BT006235.1 | - | 1 - 2 | 166 | 81 | ||
More Information
Set Membership: |
|

PRP2
Genomic Map