Assay Details
| Gene Symbol: TIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335732.1 | NP_001323967.1 | 2 - 3 | 454 | 103 | |
| NM_127687.4 | NP_179713.1 | 1 - 2 | 415 | 103 | ||
| GenBank mRNA | AF247559.1 | - | 1 - 2 | 252 | 103 | |
| AF378898.1 | - | 1 - 2 | 241 | 103 | ||
| AY052748.1 | - | 1 - 2 | 192 | 103 | ||
| AY087893.1 | - | 1 - 2 | 286 | 103 | ||
More Information
Set Membership: |
|

TIM
Genomic Map