Assay Details
| Gene Symbol: TIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084462.1 | NP_001077931.1 | 5 - 6 | 734 | 95 | |
| NM_001335732.1 | NP_001323967.1 | 7 - 8 | 896 | 95 | ||
| NM_127687.4 | NP_179713.1 | 6 - 7 | 857 | 95 | ||
| GenBank mRNA | AF247559.1 | - | 6 - 7 | 694 | 95 | |
| AF378898.1 | - | 6 - 7 | 683 | 95 | ||
| AY052748.1 | - | 6 - 7 | 634 | 95 | ||
| AY087893.1 | - | 6 - 7 | 728 | 95 | ||
More Information
Set Membership: |
|

TIM
Genomic Map