Assay Details
| Gene Symbol: TIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084462.1 | NP_001077931.1 | 6 - 7 | 912 | 85 | |
| NM_001335732.1 | NP_001323967.1 | 8 - 9 | 1074 | 85 | ||
| NM_127687.4 | NP_179713.1 | 7 - 8 | 1035 | 85 | ||
| GenBank mRNA | AF247559.1 | - | 7 - 8 | 872 | 85 | |
| AF378898.1 | - | 7 - 8 | 861 | 85 | ||
| AY052748.1 | - | 7 - 8 | 812 | 85 | ||
| AY087893.1 | - | 7 - 8 | 906 | 85 | ||
More Information
Set Membership: |
|

TIM
Genomic Map