Assay Details
| Gene Symbol: AHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113860.4 | NP_189581.1 | 1 - 2 | 211 | 107 | |
| NM_180318.2 | NP_850649.1 | 1 - 2 | 181 | 107 | ||
| GenBank mRNA | AB012568.1 | - | 1 - 2 | 168 | 107 | |
| AB015142.1 | - | 1 - 2 | 170 | 107 | ||
| AY064148.1 | - | 1 - 2 | 138 | 107 | ||
| AY097403.1 | - | 1 - 2 | 57 | 107 | ||
More Information
Set Membership: |
|

AHP2
Genomic Map