Assay Details
| Gene Symbol: AHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113860.4 | NP_189581.1 | 3 - 4 | 426 | 111 | |
| NM_180318.2 | NP_850649.1 | 3 - 4 | 396 | 111 | ||
| GenBank mRNA | AB012568.1 | - | 3 - 4 | 383 | 111 | |
| AB015142.1 | - | 3 - 4 | 385 | 111 | ||
| AY064148.1 | - | 3 - 4 | 353 | 111 | ||
| AY097403.1 | - | 3 - 4 | 272 | 111 | ||
More Information
Set Membership: |
|

AHP2
Genomic Map