Assay Details
| Gene Symbol: ASN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114602.4 | NP_190318.1 | 3 - 4 | 706 | 100 | |
| NM_180332.1 | NP_850663.1 | 3 - 4 | 517 | 100 | ||
| NM_180333.1 | NP_850664.1 | 3 - 4 | 517 | 100 | ||
| GenBank mRNA | AF419557.1 | - | 3 - 4 | 514 | 100 | |
| AY072214.1 | - | 3 - 4 | 519 | 100 | ||
| AY096592.1 | - | 3 - 4 | 318 | 100 | ||
| L29083.1 | - | 3 - 4 | 318 | 100 | ||
More Information
Set Membership: |
|

ASN1
Genomic Map