Assay Details
| Gene Symbol: ATG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203131.1 | NP_001190060.1 | 2 - 3 | 391 | 119 | |
| NM_115039.4 | NP_190748.1 | 2 - 3 | 362 | 119 | ||
| NM_180348.3 | NP_850679.1 | 2 - 3 | 324 | 119 | ||
| GenBank mRNA | AF422841.1 | - | 2 - 3 | 183 | 119 | |
| AY056616.1 | - | 2 - 3 | 279 | 119 | ||
| AY142627.1 | - | 2 - 3 | 183 | 119 | ||
| U72503.1 | - | 2 - 3 | 279 | 119 | ||
More Information
Set Membership: |
|

ATG2
Genomic Map