Assay Details
| Gene Symbol: GR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180352.3 | NP_850683.1 | 1 - 2 | 1413 | 111 | |
| GenBank mRNA | DQ925470.1 | - | 1 - 2 | 1399 | 111 | |
More Information
Set Membership: |
|

GR1
Genomic Map