Assay Details
| Gene Symbol: ACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339583.1 | NP_001319732.1 | 3 - 4 | 465 | 90 | |
| NM_001339584.1 | NP_001326182.1 | 1 - 2 | 237 | 90 | ||
| NM_115132.3 | NP_566971.2 | 2 - 3 | 1007 | 90 | ||
| NM_180354.2 | NP_850685.1 | 2 - 3 | 131 | 90 | ||
| GenBank mRNA | U73462.1 | - | 2 - 3 | 131 | 90 | |
More Information
Set Membership: |
|

ACA1
Genomic Map