Assay Details
| Gene Symbol: ACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339583.1 | NP_001319732.1 | 5 - 6 | 897 | 71 | |
| NM_001339584.1 | NP_001326182.1 | 3 - 4 | 669 | 71 | ||
| NM_115132.3 | NP_566971.2 | 4 - 5 | 1439 | 71 | ||
| NM_180354.2 | NP_850685.1 | 4 - 5 | 563 | 71 | ||
| GenBank mRNA | U73462.1 | - | 4 - 5 | 563 | 71 | |
More Information
Set Membership: |
|

ACA1
Genomic Map