Assay Details
| Gene Symbol: FC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035776.1 | NP_001030853.1 | 5 - 6 | 545 | 134 | |
| NM_001339622.1 | NP_001327608.1 | 5 - 6 | 493 | 134 | ||
| NM_115217.3 | NP_190925.1 | 4 - 5 | 564 | 134 | ||
| NM_180364.4 | NP_850695.2 | 3 - 4 | 773 | 134 | ||
| NM_202696.3 | NP_974425.1 | 4 - 5 | 518 | 134 | ||
| GenBank mRNA | AK317353.1 | - | 5 - 6 | 546 | 134 | |
| D45354.1 | - | 4 - 5 | 524 | 134 | ||
More Information
Set Membership: |
|

FC1
Genomic Map