Assay Details
| Gene Symbol: FC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035776.1 | NP_001030853.1 | 8 - 9 | 851 | 64 | |
| NM_001339622.1 | NP_001327608.1 | 8 - 9 | 799 | 64 | ||
| NM_115217.3 | NP_190925.1 | 7 - 8 | 870 | 64 | ||
| NM_180364.4 | NP_850695.2 | 6 - 7 | 1079 | 64 | ||
| NM_202696.3 | NP_974425.1 | 7 - 8 | 824 | 64 | ||
| GenBank mRNA | AK317353.1 | - | 8 - 9 | 852 | 64 | |
| D45354.1 | - | 7 - 8 | 830 | 64 | ||
More Information
Set Membership: |
|

FC1
Genomic Map