Assay Details
| Gene Symbol: FC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035776.1 | NP_001030853.1 | 9 - 10 | 948 | 111 | |
| NM_001339622.1 | NP_001327608.1 | 9 - 10 | 896 | 111 | ||
| NM_115217.3 | NP_190925.1 | 8 - 9 | 967 | 111 | ||
| NM_180364.4 | NP_850695.2 | 7 - 8 | 1176 | 111 | ||
| NM_202696.3 | NP_974425.1 | 8 - 9 | 921 | 111 | ||
| GenBank mRNA | AK317353.1 | - | 9 - 10 | 949 | 111 | |
| D45354.1 | - | 8 - 9 | 927 | 111 | ||
More Information
Set Membership: |
|

FC1
Genomic Map