Assay Details
| Gene Symbol: RBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115238.2 | NP_566989.1 | 5 - 6 | 1241 | 126 | |
| NM_180367.2 | NP_850698.1 | 4 - 5 | 1311 | 126 | ||
| GenBank mRNA | BT029533.1 | - | 3 - 4 | 561 | 126 | |
More Information
Set Membership: |
|

RBL4
Genomic Map