Assay Details
| Gene Symbol: AT3G62940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203236.2 | NP_001190165.1 | 2 - 3 | 344 | 78 | |
| NM_001340185.1 | NP_001325820.1 | 1 - 2 | 596 | 78 | ||
| NM_001340186.1 | NP_001325819.1 | 2 - 3 | 312 | 78 | ||
| NM_116159.4 | NP_191853.1 | 2 - 3 | 193 | 78 | ||
| NM_180408.3 | NP_850739.1 | 2 - 3 | 177 | 78 | ||
| GenBank mRNA | AY062840.1 | - | 2 - 3 | 144 | 78 | |
| AY081578.1 | - | 1 - 2 | 65 | 78 | ||
| AY085474.1 | - | 2 - 3 | 155 | 78 | ||
| JQ013449.1 | - | 1 - 2 | 113 | 78 | ||
| JQ013450.1 | - | 1 - 2 | 68 | 78 | ||
More Information
Set Membership: |
|

AT3G62940
Genomic Map