Assay Details
| Gene Symbol: AT3G63480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116213.3 | NP_567148.1 | 11 - 12 | 785 | 82 | |
| NM_180411.3 | NP_850742.1 | 11 - 12 | 855 | 82 | ||
| GenBank mRNA | AK119094.1 | - | 11 - 12 | 773 | 82 | |
| BT005391.1 | - | 10 - 11 | 677 | 82 | ||
More Information
Set Membership: |
|

AT3G63480
Genomic Map